Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ccdc79
Entrez Gene ID
320022
Gene Name
coiled-coil domain containing 79
Gene Aliases
4930532D21Rik, BB085179, Terb1
Chromosome Location
Chr.8: 104446718 - 104509887 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_180958.3 NP_851289.2 15 - 16 1904 119
GenBank mRNA AB775896.1 - 13 - 14 1682 119
AK029754.1 - 15 - 16 1905 119
AK030157.1 - 14 - 15 2218 119
BC113771.1 - 12 - 13 1944 119
BC129814.1 - 12 - 13 1948 119

View Full Product Details