Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Klk1b11
Entrez Gene ID
16613
Gene Name
kallikrein 1-related peptidase b11
Gene Aliases
Klk11, mGK-11
Chromosome Location
Chr.7: 43995879 - 43999875 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010640.1 NP_034770.1 - 524 129
GenBank mRNA BC013660.1 - - 524 129

View Full Product Details