The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028639.3 | NP_082915.1 | 19 - 20 | 2692 | 52 |
GenBank mRNA | AK040578.1 | - | 19 - 20 | 2695 | 52 |
AK040610.1 | - | 19 - 20 | 2663 | 52 | |
AK050263.1 | - | 19 - 20 | 2679 | 52 | |
AK080284.1 | - | 19 - 20 | 2671 | 52 | |
AK165244.1 | - | 12 - 13 | 1354 | 52 | |
BC023773.1 | - | 15 - 16 | 1650 | 52 | |
BC025613.1 | - | 1 - 2 | 175 | 52 | |
BC042512.1 | - | 19 - 20 | 2369 | 52 | |
BC049254.1 | - | 3 - 4 | 415 | 52 | |
BC057906.1 | - | 2 - 3 | 315 | 52 |