The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177696.1 | NP_001171167.1 | 1 - 2 | 184 | 73 |
NM_001177697.1 | NP_001171168.1 | 1 - 2 | 184 | 73 | |
NM_001177698.1 | NP_001171169.1 | 1 - 2 | 184 | 73 | |
NM_001177699.1 | NP_001171170.1 | 1 - 2 | 184 | 73 | |
NM_001177700.1 | NP_001171171.1 | 1 - 2 | 184 | 73 | |
NM_177678.7 | NP_808346.3 | 1 - 2 | 184 | 73 | |
GenBank mRNA | AJ748602.1 | - | 1 - 2 | 140 | 73 |
AJ748603.1 | - | 1 - 2 | 140 | 73 | |
AK046879.1 | - | 1 - 2 | 133 | 73 | |
AK082707.1 | - | 1 - 2 | 134 | 73 | |
AK154494.1 | - | 1 - 2 | 186 | 73 | |
AY274116.1 | - | 1 - 2 | 111 | 73 | |
BC141125.1 | - | 1 - 2 | 44 | 73 |