The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011155.2 | NP_035285.2 | 9 - 10 | 1237 | 118 |
GenBank mRNA | AF018262.1 | - | 9 - 10 | 1208 | 118 |
AK189512.1 | - | 2 - 3 | 121 | 118 | |
BC003744.1 | - | 9 - 10 | 1193 | 118 |