Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nr2c1
Entrez Gene ID
22025
Gene Name
nuclear receptor subfamily 2, group C, member 1
Gene Aliases
4831444H07Rik, 80.3, Eenr, TR2, Tr2-11
Chromosome Location
Chr.10: 94147517 - 94197216 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011629.3 NP_035759.3 2 - 3 241 76
GenBank mRNA AK146891.1 - 2 - 3 254 76
AK149374.1 - 2 - 3 156 76
BC090662.1 - 2 - 3 217 76
BC094580.1 - 2 - 3 159 76
Y11436.1 - 1 - 2 689 76

View Full Product Details