The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007494.3 | NP_031520.1 | 5 - 6 | 504 | 125 |
GenBank mRNA | AK146635.1 | - | 5 - 6 | 473 | 125 |
AK149489.1 | - | 5 - 6 | 476 | 125 | |
BC002074.1 | - | 5 - 6 | 504 | 125 | |
BC087556.1 | - | 4 - 5 | 433 | 125 | |
M31690.1 | - | 5 - 6 | 474 | 125 |