The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009855.2 | NP_033985.3 | 4 - 5 | 1071 | 69 |
GenBank mRNA | AF065893.1 | - | 4 - 5 | 880 | 69 |
AF065894.1 | - | 4 - 5 | 880 | 69 | |
AF065895.1 | - | 4 - 5 | 880 | 69 | |
AF065896.1 | - | 4 - 5 | 880 | 69 | |
AJ278965.1 | - | 4 - 5 | 823 | 69 | |
AY278186.1 | - | 4 - 5 | 871 | 69 | |
BC131959.1 | - | 4 - 5 | 891 | 69 | |
X60958.1 | - | 4 - 5 | 1071 | 69 |