Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Epsti1
Entrez Gene ID
108670
Gene Name
epithelial stromal interaction 1 (breast)
Gene Aliases
2310046K10Rik, 5033415K03Rik, BRESI1
Chromosome Location
Chr.14: 77904239 - 78002656 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029495.2 NP_083771.1 4 - 5 647 72
NM_178825.4 NP_849147.1 4 - 5 647 72
GenBank mRNA AK017174.1 - 4 - 5 480 72
AK077223.1 - 4 - 5 453 72
AK077376.1 - 4 - 5 482 72
AK135710.1 - 4 - 5 641 72
AK136055.1 - 4 - 5 648 72
AY239223.1 - 4 - 5 480 72
AY239224.1 - 4 - 5 553 72
BC020120.1 - 4 - 5 480 72
BC021821.1 - 4 - 5 507 72

View Full Product Details