The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029546.2 | NP_083822.1 | 7 - 8 | 899 | 81 |
GenBank mRNA | AB041855.1 | - | 7 - 8 | 860 | 81 |
AK088254.1 | - | 7 - 8 | 900 | 81 | |
AK088983.1 | - | 7 - 8 | 900 | 81 | |
BC112325.1 | - | 7 - 8 | 865 | 81 | |
BC128480.1 | - | 7 - 8 | 880 | 81 |