Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sun5
Entrez Gene ID
76407
Gene Name
Sad1 and UNC84 domain containing 5
Gene Aliases
1700021O15Rik, SPAG4L-2, SRG4, Spag4l
Chromosome Location
Chr.2: 153856188 - 153871095 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK006225.1 - 3 - 4 268 71
AY307077.1 - 3 - 4 287 71
FJ667498.1 - 4 - 5 276 71

View Full Product Details