Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Catsper3
Entrez Gene ID
76856
Gene Name
cation channel, sperm associated 3
Gene Aliases
4921522D01Rik, CACRC
Chromosome Location
Chr.13: 55784579 - 55824942 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252487.1 NP_001239416.1 4 - 5 756 70
NM_001252488.1 NP_001239417.1 4 - 5 603 70
NM_029772.4 NP_084048.1 4 - 5 717 70
GenBank mRNA AK014942.1 - 4 - 5 718 70
AY263400.1 - 4 - 5 731 70
BC089518.1 - - 856 70
HM179540.1 - 4 - 5 673 70
HM179541.1 - 4 - 5 520 70

View Full Product Details