The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029879.2 | NP_084155.2 | 1 - 2 | 885 | 75 |
GenBank mRNA | AK020910.1 | - | 1 - 2 | 482 | 75 |
AK028323.1 | - | 1 - 2 | 262 | 75 | |
AK032138.1 | - | 1 - 2 | 885 | 75 | |
AK036240.1 | - | 1 - 2 | 792 | 75 | |
AK046733.1 | - | 1 - 2 | 514 | 75 | |
AK137417.1 | - | 1 - 2 | 759 | 75 | |
BC118957.2 | - | 1 - 2 | 417 | 75 | |
DQ104214.1 | - | 1 - 2 | 175 | 75 |