The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029949.3 | NP_084225.1 | 7 - 8 | 983 | 102 |
GenBank mRNA | AK007563.1 | - | 7 - 8 | 983 | 102 |
AK019863.2 | - | 7 - 8 | 979 | 102 | |
AK031870.1 | - | 7 - 8 | 985 | 102 | |
AK031924.1 | - | 6 - 7 | 901 | 102 | |
AK049933.1 | - | 7 - 8 | 1004 | 102 | |
BC048713.1 | - | 7 - 8 | 983 | 102 |