The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133786.3 | NP_598547.1 | 2 - 3 | 256 | 119 |
GenBank mRNA | AJ534940.2 | - | 1 - 2 | 132 | 119 |
AK088350.1 | - | 2 - 3 | 239 | 119 | |
AK088846.1 | - | 2 - 3 | 222 | 119 | |
AK133817.1 | - | 2 - 3 | 246 | 119 | |
AK152804.1 | - | 2 - 3 | 283 | 119 | |
BC062939.1 | - | 2 - 3 | 256 | 119 |