The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173765.3 | NP_776126.1 | 15 - 16 | 3203 | 57 |
GenBank mRNA | AB095954.1 | - | 15 - 16 | 3220 | 57 |
AK136849.1 | - | 15 - 16 | 3203 | 57 | |
AK148405.1 | - | 14 - 15 | 3029 | 57 | |
BC128330.1 | - | 14 - 15 | 3075 | 57 |