The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175022.2 | NP_778187.2 | 9 - 10 | 5495 | 135 |
GenBank mRNA | AK032894.1 | - | 6 - 7 | 1983 | 135 |
AK173130.1 | - | 6 - 7 | 4934 | 135 | |
BC025846.1 | - | 1 - 2 | 656 | 135 | |
BC058674.1 | - | 6 - 7 | 4134 | 135 | |
BC075628.1 | - | 10 - 11 | 4405 | 135 |