The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083810.2 | NP_001077279.1 | 8 - 9 | 1109 | 86 |
NM_001110849.1 | NP_001104319.1 | 9 - 10 | 972 | 86 | |
NM_175181.5 | NP_780390.3 | 8 - 9 | 1058 | 86 | |
GenBank mRNA | AK011178.1 | - | 3 - 4 | 306 | 86 |
AK030499.1 | - | 8 - 9 | 1016 | 86 | |
AK145981.1 | - | 8 - 9 | 1060 | 86 | |
BC048394.1 | - | 8 - 9 | 968 | 86 | |
BC172132.1 | - | 7 - 8 | 784 | 86 | |
BC172158.1 | - | 6 - 7 | 692 | 86 |