The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175214.3 | NP_780423.2 | 13 - 14 | 3155 | 82 |
GenBank mRNA | AK032912.1 | - | 6 - 7 | 1010 | 82 |
BC116646.1 | - | 11 - 12 | 1994 | 82 |