The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175563.5 | NP_780772.1 | 2 - 3 | 208 | 106 |
GenBank mRNA | AK040348.1 | - | 2 - 3 | 186 | 106 |
AK047463.1 | - | 2 - 3 | 186 | 106 | |
AK083651.1 | - | 2 - 3 | 203 | 106 | |
BC023918.1 | - | 2 - 3 | 218 | 106 |