The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177338.5 | NP_796312.2 | 3 - 4 | 823 | 129 |
GenBank mRNA | AK052729.1 | - | 3 - 4 | 833 | 129 |
AK089782.1 | - | 3 - 4 | 824 | 129 | |
AK161592.1 | - | 3 - 4 | 823 | 129 | |
BC002212.1 | - | 1 - 2 | 227 | 129 | |
BC051457.1 | - | 3 - 4 | 826 | 129 |