The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177338.5 | NP_796312.2 | 9 - 10 | 1447 | 66 |
GenBank mRNA | AK089782.1 | - | 9 - 10 | 1448 | 66 |