The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177778.4 | NP_808446.1 | 2 - 3 | 492 | 63 |
GenBank mRNA | AK042345.1 | - | 2 - 3 | 328 | 63 |
AK146249.1 | - | 2 - 3 | 454 | 63 | |
BC023126.1 | - | 2 - 3 | 492 | 63 |