Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rnf24
Entrez Gene ID
51902
Gene Name
ring finger protein 24
Gene Aliases
2810473M14Rik, 4930505A13Rik, AI317164, C86507, D2Ertd504e
Chromosome Location
Chr.2: 131298488 - 131352892 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178607.4 NP_848722.1 4 - 5 398 140
GenBank mRNA AK048813.1 - 4 - 5 356 140
AK083665.1 - 4 - 5 388 140
AK089207.1 - 4 - 5 385 140
AK154093.1 - 4 - 5 335 140
AK165427.1 - 4 - 5 356 140
BC096529.1 - 4 - 5 331 140

View Full Product Details