Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rnf24
Entrez Gene ID
51902
Gene Name
ring finger protein 24
Gene Aliases
2810473M14Rik, 4930505A13Rik, AI317164, C86507, D2Ertd504e
Chromosome Location
Chr.2: 131298488 - 131352892 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178607.4 NP_848722.1 5 - 6 470 78
GenBank mRNA AK015699.1 - 6 - 7 530 78
AK048813.1 - 5 - 6 428 78
AK083665.1 - 5 - 6 460 78
AK089207.1 - 5 - 6 457 78
AK165427.1 - 5 - 6 428 78
BC096529.1 - 5 - 6 403 78

View Full Product Details