The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178633.3 | NP_848748.2 | - | 1877 | 80 |
GenBank mRNA | AK020050.1 | - | - | 528 | 80 |
AK032116.1 | - | - | 1425 | 80 | |
AK167903.1 | - | - | 1668 | 80 | |
AK220570.1 | - | - | 1877 | 80 | |
BC031142.1 | - | - | 1748 | 80 | |
BC031144.1 | - | - | 1771 | 80 |