The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199247.1 | NP_001186176.1 | 5 - 6 | 432 | 93 |
NM_001199248.1 | NP_001186177.1 | 4 - 5 | 531 | 93 | |
NM_001199249.1 | NP_001186178.1 | 4 - 5 | 531 | 93 | |
NM_178637.2 | NP_848752.1 | 6 - 7 | 588 | 93 | |
NR_037603.1 | 3 - 4 | 464 | 93 | ||
GenBank mRNA | AB055409.1 | - | 3 - 4 | 442 | 93 |
AF528194.1 | - | 2 - 3 | 142 | 93 | |
AF528195.1 | - | 5 - 6 | 523 | 93 | |
AF528196.1 | - | 4 - 5 | 367 | 93 | |
AK143674.1 | - | 4 - 5 | 483 | 93 | |
AK146352.1 | - | 6 - 7 | 545 | 93 | |
AK159568.1 | - | 4 - 5 | 464 | 93 | |
AK160077.1 | - | 4 - 5 | 464 | 93 | |
AK206382.1 | - | 1 - 2 | 91 | 93 | |
BC110675.1 | - | 6 - 7 | 529 | 93 | |
BC129967.1 | - | 4 - 5 | 366 | 93 | |
BC129968.1 | - | 4 - 5 | 367 | 93 | |
DQ139980.1 | - | 5 - 6 | 523 | 93 |