The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114660.2 | NP_001108132.1 | 1 - 2 | 959 | 66 |
NM_001286437.1 | NP_001273366.1 | 1 - 2 | 959 | 66 | |
NM_178672.7 | NP_848787.2 | 1 - 2 | 959 | 66 | |
GenBank mRNA | AK034000.1 | - | 1 - 2 | 954 | 66 |
AK036637.1 | - | 1 - 2 | 901 | 66 | |
AK044408.1 | - | 1 - 2 | 938 | 66 | |
AK047026.1 | - | 1 - 2 | 595 | 66 | |
AK047738.1 | - | 1 - 2 | 902 | 66 | |
AK050673.1 | - | 1 - 2 | 910 | 66 | |
AK054548.1 | - | 1 - 2 | 918 | 66 | |
AK088353.1 | - | 1 - 2 | 845 | 66 | |
AK089011.1 | - | 1 - 2 | 660 | 66 | |
AK089153.2 | - | 2 - 3 | 953 | 66 | |
AK135666.1 | - | 1 - 2 | 195 | 66 | |
AK140246.1 | - | 1 - 2 | 960 | 66 | |
AK143615.1 | - | 1 - 2 | 939 | 66 | |
AK153621.1 | - | 1 - 2 | 960 | 66 | |
AK163028.1 | - | 1 - 2 | 918 | 66 | |
BC042528.1 | - | 1 - 2 | 970 | 66 |