Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Egflam
Entrez Gene ID
268780
Gene Name
EGF-like, fibronectin type III and laminin G domains
Gene Aliases
5930412K08, AU040377
Chromosome Location
Chr.15: 7206120 - 7398746 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289496.1 NP_001276425.1 21 - 22 3102 99
NM_001289498.1 NP_001276427.1 21 - 22 3097 99
NM_178748.6 NP_848863.1 20 - 21 3078 99
GenBank mRNA AK031136.1 - 20 - 21 2987 99
AK033332.1 - 20 - 21 2995 99
AK037223.1 - 20 - 21 2980 99
AK041546.1 - 20 - 21 2959 99
AK158909.1 - 21 - 22 3098 99
BC051455.1 - 6 - 7 631 99
BC095994.1 - 21 - 22 2832 99
BC150710.1 - 20 - 21 2798 99
DQ223720.1 - 21 - 22 2806 99

View Full Product Details