The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178883.5 | NP_849214.2 | 2 - 3 | 471 | 65 |
GenBank mRNA | AK043953.1 | - | 2 - 3 | 472 | 65 |
AK165909.1 | - | 2 - 3 | 441 | 65 | |
AK169151.1 | - | 2 - 3 | 464 | 65 | |
BC119132.1 | - | 2 - 3 | 454 | 65 | |
BC119134.1 | - | 2 - 3 | 454 | 65 |