The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289572.1 | NP_001276501.1 | 10 - 11 | 1254 | 79 |
NM_001289573.1 | NP_001276502.1 | 9 - 10 | 849 | 79 | |
NM_001289574.1 | NP_001276503.1 | 11 - 12 | 1416 | 79 | |
NM_178914.4 | NP_849245.2 | 11 - 12 | 1350 | 79 | |
GenBank mRNA | AK036359.1 | - | 11 - 12 | 1334 | 79 |
AK077500.1 | - | 11 - 12 | 1261 | 79 | |
AK153698.1 | - | 9 - 10 | 825 | 79 | |
AK161308.1 | - | 10 - 11 | 1200 | 79 | |
BC046960.1 | - | 11 - 12 | 1308 | 79 | |
DQ196101.1 | - | 10 - 11 | 1080 | 79 |