The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181579.1 | NP_853557.1 | 9 - 10 | 1063 | 97 |
GenBank mRNA | AF309775.1 | - | 8 - 9 | 956 | 97 |
AK086511.1 | - | 9 - 10 | 1064 | 97 |