Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sirt6
Entrez Gene ID
50721
Gene Name
sirtuin 6
Gene Aliases
2810449N18Rik, AI043036, Sir2l6
Chromosome Location
Chr.10: 81621786 - 81627792 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181586.3 NP_853617.1 1 - 2 136 72
GenBank mRNA AK013316.1 - 1 - 2 85 72
BC052763.1 - 1 - 2 86 72

View Full Product Details