Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccdc67
Entrez Gene ID
234964
Gene Name
coiled-coil domain containing 67
Gene Aliases
4933401K09Rik, Deup1
Chromosome Location
Chr.9: 15559864 - 15628338 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181816.2 NP_861537.2 6 - 7 698 85
GenBank mRNA AK030153.1 - 6 - 7 681 85
AK077147.1 - 1 - 2 76 85
BC096547.1 - 5 - 6 591 85
KC211186.1 - 5 - 6 542 85

View Full Product Details