Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rerg
Entrez Gene ID
232441
Gene Name
RAS-like, estrogen-regulated, growth-inhibitor
Gene Aliases
-
Chromosome Location
Chr.6: 137054817 - 137170496 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164212.1 NP_001157684.1 2 - 3 406 72
NM_181988.2 NP_871788.1 2 - 3 226 72
GenBank mRNA AK133734.1 - 2 - 3 388 72
BC026463.1 - 2 - 3 239 72

View Full Product Details