The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144953.1 | NP_001138425.1 | 6 - 7 | 797 | 69 |
NM_182636.4 | NP_872574.2 | 6 - 7 | 797 | 69 | |
GenBank mRNA | AK141799.1 | - | 6 - 7 | 798 | 69 |
BC052055.1 | - | 5 - 6 | 743 | 69 |