The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007917.3 | NP_031943.3 | 8 - 8 | 2669 | 138 |
GenBank mRNA | AK082867.1 | - | 7 - 7 | 2550 | 138 |