Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sprr2i
Entrez Gene ID
20763
Gene Name
small proline-rich protein 2I
Gene Aliases
-
Chromosome Location
Chr.3: 92407991 - 92409271 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011475.3 NP_035605.1 2 - 2 348 73
GenBank mRNA AY158993.1 - 2 - 2 348 73

View Full Product Details