The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019822.3 | NP_062796.2 | 2 - 2 | 139 | 79 |
GenBank mRNA | AF225959.1 | - | 2 - 2 | 55 | 79 |
AK010919.1 | - | 2 - 2 | 116 | 79 | |
AK082084.1 | - | 2 - 2 | 122 | 79 | |
AK143843.1 | - | 2 - 2 | 114 | 79 | |
AK145474.1 | - | 2 - 2 | 101 | 79 | |
AK145792.1 | - | 2 - 2 | 130 | 79 | |
AK150091.1 | - | 2 - 2 | 91 | 80 | |
AK166878.1 | - | 2 - 2 | 128 | 79 | |
BC008974.1 | - | 2 - 2 | 139 | 79 | |
BC019746.1 | - | 2 - 2 | 68 | 79 | |
BC031517.1 | - | 2 - 2 | 82 | 79 |