The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204875.1 | NP_001191804.1 | 7 - 7 | 1074 | 153 |
NM_023871.4 | NP_076360.1 | 7 - 7 | 939 | 153 | |
GenBank mRNA | AB044937.1 | - | 7 - 7 | 823 | 153 |
AK011630.1 | - | 5 - 5 | 336 | 153 | |
AK019960.1 | - | 7 - 7 | 823 | 153 | |
AK172327.1 | - | 7 - 7 | 1083 | 153 | |
BC018255.1 | - | 7 - 7 | 892 | 153 | |
BC103791.1 | - | 7 - 7 | 767 | 153 |