The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025887.4 | NP_080163.1 | 6 - 6 | 1272 | 142 |
GenBank mRNA | AK002631.1 | - | 6 - 6 | 1027 | 142 |
AK008198.1 | - | 6 - 6 | 1013 | 142 | |
AK010495.1 | - | 6 - 6 | 1117 | 142 | |
AK076093.1 | - | 6 - 6 | 1252 | 142 | |
AK081999.1 | - | 6 - 6 | 1273 | 142 | |
AK150296.1 | - | 6 - 6 | 1026 | 142 | |
AK150346.1 | - | 6 - 6 | 1026 | 142 | |
AK150405.1 | - | 6 - 6 | 1070 | 142 | |
AK151066.1 | - | 6 - 6 | 1026 | 142 | |
AK153131.1 | - | 6 - 6 | 1026 | 142 | |
AK159706.1 | - | 6 - 6 | 142 | ||
BC004842.1 | - | 6 - 6 | 142 | ||
BC034370.1 | - | 6 - 6 | 142 | ||
BC096481.1 | - | 7 - 7 | 142 |