The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010005.3 | NP_034135.2 | 4 - 5 | 752 | 129 |
GenBank mRNA | BC010989.1 | - | 4 - 5 | 750 | 129 |
BC051039.1 | - | 4 - 5 | 729 | 129 | |
BC057924.1 | - | 4 - 5 | 739 | 129 | |
BF660186.1 | - | 2 - 3 | 188 | 129 | |
M27167.1 | - | 4 - 5 | 730 | 129 |