The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011966.3 | NP_036096.1 | 5 - 6 | 355 | 148 |
GenBank mRNA | AF060093.1 | - | 5 - 6 | 340 | 148 |
AK085937.1 | - | 5 - 6 | 413 | 148 | |
AK150565.1 | - | 5 - 6 | 400 | 148 | |
AK166618.1 | - | 5 - 6 | 394 | 148 | |
AK168150.1 | - | 5 - 6 | 371 | 148 | |
AK168838.1 | - | 5 - 6 | 414 | 148 | |
BC001982.1 | - | 5 - 6 | 355 | 148 |