Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pogk
Entrez Gene ID
71592
Gene Name
pogo transposable element with KRAB domain
Gene Aliases
9130401E23Rik, BASS2, mKIAA1513
Chromosome Location
Chr.1: 166393610 - 166409858 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175170.4 NP_780379.2 1 - 2 123 70
GenBank mRNA AK031716.1 - 1 - 2 76 70
AK048067.1 - 1 - 2 82 70
AK053898.1 - 1 - 2 101 70
CJ102323.1 - 1 - 2 123 70

View Full Product Details