The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178691.4 | NP_848806.2 | 1 - 2 | 489 | 78 |
GenBank mRNA | AK036938.1 | - | 1 - 2 | 490 | 78 |
AK036991.1 | - | 1 - 2 | 491 | 78 | |
AK044678.1 | - | 1 - 2 | 284 | 78 | |
AK053730.1 | - | 1 - 2 | 279 | 78 | |
BC099948.1 | - | 1 - 2 | 282 | 78 | |
BC139034.1 | - | 1 - 2 | 415 | 78 |