The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177371.3 | NP_796345.3 | 3 - 4 | 383 | 113 |
GenBank mRNA | AF520786.1 | - | 3 - 4 | 300 | 113 |
AY764130.1 | - | 3 - 4 | 383 | 113 | |
BC096494.1 | - | 3 - 4 | 369 | 113 | |
BC138207.1 | - | 3 - 4 | 348 | 113 | |
BC138208.1 | - | 3 - 4 | 348 | 113 |