Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Copb1
Entrez Gene ID
70349
Gene Name
coatomer protein complex, subunit beta 1
Gene Aliases
2610019B04Rik
Chromosome Location
Chr.7: 114215559 - 114254680 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033370.3 NP_203534.1 6 - 7 835 133
GenBank mRNA AF231925.1 - 5 - 6 774 133
AK135168.1 - 6 - 7 132
AK149828.1 - 6 - 7 853 133
AK152222.1 - 6 - 7 862 133
AK161544.1 - 6 - 7 862 133
AK172214.1 - 6 - 7 837 133
BC030837.1 - 6 - 7 835 133

View Full Product Details