The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019980.2 | NP_064364.1 | 3 - 4 | 463 | 60 |
GenBank mRNA | AF171100.1 | - | 3 - 4 | 431 | 60 |
AF230522.1 | - | 2 - 3 | 413 | 60 | |
AK018578.1 | - | 3 - 4 | 446 | 60 | |
AK076162.1 | - | 3 - 4 | 465 | 60 | |
AK151053.1 | - | 3 - 4 | 463 | 60 | |
AK159533.1 | - | 3 - 4 | 431 | 60 | |
BC018559.1 | - | 3 - 4 | 443 | 60 |