The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175246.4 | NP_780455.2 | 1 - 2 | 262 | 152 |
GenBank mRNA | AK010794.1 | - | 1 - 2 | 263 | 152 |
AK040571.1 | - | 1 - 2 | 263 | 152 | |
AK049254.1 | - | 1 - 2 | 263 | 152 | |
AK049318.1 | - | 1 - 2 | 263 | 152 | |
AK132768.1 | - | 1 - 2 | 240 | 152 | |
AK143801.1 | - | 1 - 2 | 262 | 152 | |
BC064067.1 | - | 1 - 2 | 235 | 152 |