The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177093.3 | NP_796067.2 | 3 - 4 | 989 | 147 |
GenBank mRNA | AK049274.1 | - | 3 - 4 | 990 | 147 |
AK090204.1 | - | 3 - 4 | 998 | 147 | |
AK147818.1 | - | 3 - 4 | 965 | 147 | |
BC141007.1 | - | 3 - 4 | 949 | 147 |